missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine IFNG Protein
A cDNA sequence encoding the IFNG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10677-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
403801 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by an anti-viral assay using A-72 canine fibroma cells infected with vesicular stomatitis virus (VSV) is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Canine | |
Untagged | |
The IFNG was lyophilized from a 0.2microm filtered concentrated solution in 25mM Sodium Succinate, pH 5.0, 60mM NaCl, with 0.1% Tween-80. |
IFNG Protein | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
QAMFFKEIEN LKEYFNASNP DVSDGGSLFV DILKKWREES DKTIIQSQIV SFYLKLFDNF KDNQIIQRSM DTIKEDMLGK FLNSSTSKRE DFLKLIQIPV NDLQVQRKAI NELIKVMNDL SPRSNLRKRK RSQNLFRGRR ASK | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |