missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum clpP Protein
A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) clpP, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7429-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_003357557.1 | |
clpP Protein | |
Research Use Only | |
clpP | |
Recombinant Protein | |
E. coli | |
MSLVPVVVEQ TNRGERSYDI YSRLLKDRII MLSEEVNDTT ASLIVAQLLF LEAEDPDKDI HLYINSPGGS ITSGMAIYDT MQYIKPDVST ICVGMAASMG AFLLAAGAKG KRYALPNSEV MIHQPLGGFR GQATDIGIHA ERILKMKKKL NTILSDRTGK PLEQVELDTE RDHFLSAEEA KEYGLIDEVI DKKK | |
Greater than 90% as determined by SDS-PAGE. |
5187372 | |
500 μg | |
Not determined. | |
Full Length | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |