missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type D Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type D, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9546-ye-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Botulinum neurotoxin type D Protein | |
Research Use Only | |
botD | |
Recombinant Protein | |
Yeast | |
MTWPVKDFNY SDPVNDNDIL YLRIPQNKLI TTPVKAFMIT QNIWVIPERF SSDTNPSLSK PPRPTSKYQS YYDPSYLSTD EQKDTFLKGI IKLFKRINER DIGKKLINYL VVGSPFMGDS STPEDTFDFT RHTTNIAVEK FENGSWKVTN IITPSVLIFG PLPNILDYTA SLTLQGQQSN PSFEGFGTLS ILKVAPEFLL TFSDVTSNQS SAVLGKSIFC MDPVIALMHE LTHSLHQLYG INIPSDKRIR PQVSEGFFSQ DGPNVQFEEL YTFGGLDVEI IPQIERSQLR EKALGHYKDI AKRLNNINKT IPSSWISNID KYKKIFSEKY NFDKDNTGNF VVNIDKFNSL YSDLTNVMSE VVYSSQYNVK NRTHYFSRHY LPVFANILDD NIYTIRDGFN LTNKGFNIEN SGQNIERNPA LQKLSSESVV DLFTKVCLRL TK | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
C. botulinum | |
His | |
Tris-based buffer, 50% glycerol |