missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine TOP1 Protein
A cDNA sequence encoding the TOP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP13784-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
TOP1 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
TOP1 is supplied at a concentration of 1 mg/1ml in 16mM HEPES buffer pH 7.5, 400mM NaCl and 20% glycerol. |
Greater than 70% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Native Protein | |
DAKVLKDAKT KKVVESKKKA VQRLEEQLMK LEVQATDREE NKQIALGTSK LNYLDPRITV AWCKKWG |