missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Pregnancy-associated glycoprotein 1
A DNA sequence encoding the Bos taurus (Bovine) Pregnancy-associated glycoprotein 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7639-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_776836.1 | |
Pregnancy-associated glycoprotein 1 | |
Research Use Only | |
PAG1 | |
Recombinant Protein | |
E. coli | |
RGSNLTTHPL RNIKDLVYMG NITIGTPPQE FQVVFDTASS DLWVPSDFCT SPACSTHVRF RHLQSSTFRL TNKTFRITYG SGRMKGVVVH DTVRIGNLVS TDQPFGLSIE EYGFEGRIYD GVLGLNYPNI SFSGAIPIFD KLKNQRAISE PVFAFYLSKD EREGSVVMFG GVDHRYYEGE LNWVPLIQAG DWSVHMDRIS IERKIIACSD GCKALVDTGT SDIVGPRRLV NNIHRLIGAI PRGSEHYVPC SEVNTLPSIV FTINGINYPV PGRAYILKDD RGRCYTTFQE NRVSSSTETW YLGDVFLRLY FSVFDRGNDR IGLARAV | |
Greater than 88.1% as determined by SDS-PAGE. |
281964 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |