missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine NucleoProtein
A DNA sequence encoding the Bos taurus (Bovine) Nucleoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7198-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NucleoProtein | |
Research Use Only | |
N | |
Recombinant Protein | |
E. coli | |
MALSKVKLND TFNKDQLLST SKYTIQRSTG DNIDIPNYDV QKHLNKLCGM LLITEDANHK FTGLIGMLYA MSRLGREDTL KILKDAGYQV RANGVDVITH RQDVNGKEMK FEVLTLVSLT SEVQGNIEIE SRKSYKKMLK EMGEVAPEYR HDFPDCGMIV LCVAALVITK LAAGDRSGLT AVIRRANNVL RNEMKRYKGL IPKDIANSFY EVFEKYPHYI DVFVHFGIAQ SSTRGGSRVE GIFAGLFMNA YGAGQVMLRW GVLAKSVKNI MLGHASVQAE MEQVVEVYEY AQKLGGEAGF YHILNNPKAS LLSLTQFPNF SSVVLGNAAG LGIMGEYRGT PRNQDLYDAA KAYAEQLKEN GVINYSVLDL TTEELEAIKN QLNPKDNDVE L | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |