missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Muellerian-inhibiting factor Protein
A DNA sequence encoding the Bos taurus (Bovine) Muellerian-inhibiting factor, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8973-ye-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_776315.1 | |
Muellerian-inhibiting factor Protein | |
Research Use Only | |
AMH | |
Recombinant Protein | |
Yeast | |
REEVFSTSAL PREQATGSGA LIFQQAWDWP LSSLWLPGSP LDPLCLVTLH GSGNGSRAPL RVVGVLSSYE QAFLEAVRRT HWGLSDLTTF AVCPAGNGQP VLPHLQRLQA WLGEPGGRWL VVLHLEEVTW EPTPLLRFQE PPPGGASPPE LALLVVYPGP GLEVTVTGAG LPGTQSLCLT ADSDFLALVV DHPEGAWRRP GLALTLRRRG NGALLSTAQL QALLFGADSR CFTRKTPALL LLLPARSSAP MPAHGRLDLV PFPQPRASPE PEEAPPSADP FLETLTRLVR ALAGPPARAS PPRLALDPGA LAGFPQGQVN LSDPAALERL LDGEEPLLLL LPPTAATTGV PATPQGPKSP LWAAGLARRV AAELMS-QAV AAELRALPGL PPAAPPLLAR LLALCPGNPD SPGGPLRALL LLKALQGLRA EWRGRERSGS ARAQRSAGAA AADGPCALRE LSVDLRAERS VLIPETYQAN NCQGACGWPQ SDRNPRYGNH VVLLLKMQAR GATLARPPCC VPTAYTGKLL ISLSEERISA HHVPNMVATE CGCR | |
Greater than 90% as determined by SDS-PAGE. |
280718 | |
500 μg | |
Not determined. | |
Full Length of Mature Protein | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |