missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Inhibin alpha chain Protein
A DNA sequence encoding the Bos taurus (Bovine) Inhibin alpha chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6231-ec-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
NP_776519.2 | |
Inhibin alpha chain Protein | |
Research Use Only | |
INHA | |
Recombinant Protein | |
E. coli | |
STPPLPWPWS PAALRLLQRP PEEPAAHADC HRAALNISFQ ELGWDRWIVH PPSFIFYYCH GGCGLSPPQD LPLPVPGVPP TPVQPLSLVP GAQPCCAALP GTMRPLHVRT TSDGGYSFKY EMVPNLLTQH CACI | |
Greater than 90% as determined by SDS-PAGE. |
281254 | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |