missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine FGF 21 Protein
A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11870-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FGF 21 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (0.8 mg/ml) solution with 0.4 mg/ml of NaHCO3, pH 8. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by Gel Filtration. (b) Analysis by SDS-PAGE. |