missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Acetylcholinesterase / ACHE Protein
A DNA sequence encoding the Bos taurus (Bovine) Acetylcholinesterase / ACHE, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5606-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001069688.1 | |
Acetylcholinesterase / ACHE Protein | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
GST | |
Tris-based buffer, 50% glycerol |
540446 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ACHE | |
Recombinant Protein | |
E. coli | |
EGPEDPELLV MVRGGRLRGL RLMAPRGPVS AFLGIPFAEP PVGPRRFLPP EPKRPWPGVL NATAFQSVCY QYVDTLYPGF EGTEMWNPNR ELSEDCLYLN VWTPYPRPSS PTPVLVWIYG GGFYSGASSL DVYDGRFLTQ AEGTVLVSMN YRVGAFGFLA LPGSREAPGN VGLLDQRLAL QWVQENVAAF GGDPTSVTLF GESAGAASVG MHLLSPPSRG LFHRAVLQSG APNGPWATVG VGEARRRATL LARLVGCPPG GAGGNDTELV ACLRARPAQD LVDHEWRVLP QESVFRFSFV PVVDGDFLSD TPEALINAGD FHGLQVLVGV VKDEGSYFLV YGAPGFSKDN ESLISRAQFL AGVRVGVPQA SDLAAEAVVL HYTDWLHPED PARLREALSD VVGDHNVVCP VAQLAGRLAA QGARVYAYIF EHRASTLSWP LWMGVPHGYE IEFIFGLPLE PSLNYTIEER TFAQRLMRYW ANFARTGDPN DPRDPKAPQW PPYTAGAQQY VSLNLRPLEV RRGLRAQACA FWNRFLPKLL SATDTLDEAE RQWKAEFHRW SSYMVHWKNQ FDHYSKQDRC SDL |