missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacillus Levanase Protein
A DNA sequence encoding the Bacillus sp. (strain L7) Levanase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7478-ye-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Levanase Protein | |
Research Use Only | |
Partial (see sequence information for more details). | |
Bacillus | |
His | |
Tris-based buffer, 50% glycerol |
10 μg | |
Not determined. | |
Recombinant Protein | |
Yeast | |
LPWNDLGHVW SGSAVADTTN ASGLFGSSGG KGLIAYYTSY NPDRHNGNQK IGLAYSTDRG RTWKYSEEHP VVIENPGKTG EDPGGWDFRD PKVVRDEANN RWVMVVSGGD HIRLFTSTNL LNWTLTDQF | |
Greater than 90% as determined by SDS-PAGE. |