missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABL2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11588-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The RABL2B protein is a member of the RAB gene family which belongs to the RAS GTPase superfamily. RABL2B is located within a subtelomeric region of 22q13.3. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene.Especificaciones
RABL2B | |
Polyclonal | |
Unconjugated | |
RABL2B | |
Rab like protein 2B, RABL2B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11158 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UNT1 | |
RABL2B | |
RABL2B Fusion Protein Ag2189 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |