missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
MC4R Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MC4R.
Marca: Abnova PAB16756.100ug
                        
                            
                        
                        
                        
                        
                        
                        
                            
                                    
                                Este artículo no se puede devolver.
                                Vea la política de devoluciones
                                
                        
                    
                Especificaciones
| MC4R | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MC4R. | |
| In PBS (0.02% sodium azide) | |
| MGC126851/MGC138197 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid | 
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MC4R | |
| MC4R | |
| A synthetic peptide corresponding to N-terminus 19 amino acids of human MC4R. | |
| RUO | |
| 4160 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.  | 
                                                                    
                                                                
                    Corrección del contenido de un producto
                
                Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
			MC4R Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™ > 100μg; Unlabeled
            For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
            
                                                    
                                                    
                                                    
                            
                            
