missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IP/Rabin8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12321-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The specific function of this protein remains unknown.Especificaciones
RAB3IP/Rabin8 | |
Polyclonal | |
Unconjugated | |
RAB3IP | |
Rab 3A interacting protein, Rab3A interacting protein, RAB3IP, Rabin 3, RABIN3, Rabin8, SSX2 interacting protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117177, 216363 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q68EF0, Q96QF0 | |
Rab3ip | |
RAB3IP/Rabin8 Fusion Protein Ag2974 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |