Learn More
Estimadores de agua QuickFit™
Los estimadores de agua QuickFit™ se utilizan para determinar el contenido de agua en los productos derivados del petróleo.
Marca: Quickfit WE6/22/10T
Detalles adicionales : Peso : 0.35000kg
Descripción
- Complies with BS 756
- Dean and Stark pattern receivers for the determination of the water content in petroleum products
- For use with solvent entrainers which are lighter than water
- Borosilicate glass with graduated tube
- Receivers WE6/22/10T and WE6/33/10T are all fitted with PTFE GP Rotaflo™ stopcock for easy drainage
Especificaciones
Water Estimators | |
Borosilicate Glass | |
10 mL | |
19/26 socket |