missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PYCR1 Mouse anti-Human, Mouse, Rat, Clone: 1H2G1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66510-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.Especificaciones
PYCR1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P32322, Q922W5 | |
Pycr1 | |
PYCR1 Fusion Protein Ag17919 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG2a |
Western Blot | |
1H2G1 | |
Unconjugated | |
PYCR1 | |
P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PYCR, PYCR1 | |
Mouse | |
Protein A | |
RUO | |
209027, 287877, 5831 | |
-20°C | |
Liquid |