missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTGES2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10881-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a membrane-associated prostaglandin E synthase, which catalyzes the conversion of prostaglandin H2 to prostaglandin E2. This protein also has been shown to activate the transcription regulated by a gamma-interferon-activated transcription element (GATE). Multiple transcript variants have been found for this gene.Especificaciones
PTGES2 | |
Polyclonal | |
Unconjugated | |
PTGES2 | |
C9orf15, GBF1, mPGES 2, PGES2, prostaglandin E synthase 2, PTGES2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
311865, 80142, 96979 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8BWM0, Q9H7Z7 | |
PTGES2 | |
PTGES2 Fusion Protein Ag1329 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |