missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11029-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the OTU (ovarian tumor) domain-containing cysteine protease superfamily. The OTU domain confers deubiquitinase activity and the encoded protein has been shown to suppress the type I interferon-dependent innate immune response by cleaving the polyubiquitin chain from an essential type I interferon adaptor protein. Cleavage results in disassociation of the adaptor protein from a downstream signaling complex and disruption of the type I interferon signaling cascade. Alternatively spliced transcript variants encoding different isoforms have been described.Especificaciones
PSMB4 | |
Polyclonal | |
Unconjugated | |
PSMB4 | |
20S proteasome, 26 kDa prosomal protein, HN3, HsBPROS26, HsN3, Macropain beta chain, PROS 26, PROS26, Proteasome beta chain, Proteasome chain 3, Proteasome subunit beta type 4, PSMB4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5692 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P28070 | |
PSMB4 | |
PSMB4 Fusion Protein Ag1500 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |