missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMA2 (Human) Recombinant Protein
Human PSMA2 full-length ORF ( AAH47697, 1 a.a. - 234 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005683-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAERGYSFSLTTFSPSGKLVQIEYALAAVAGGAPSVGIKAANGVVLATEKKQKSILYDERSVHKVEPITKHIGLVYSGMGPDYRVLVHRARKLAQQYYLVYQEPIPTAQLVQRVASVMQEYTQSGGVRPFGVSLLICGWNEGRPYLFQSDPSGAYFAWKATAMGKNYVNGKTFLEKRYNEDLELEDAIHTAILTLKESFEGQMTEDNIEVGICNEAGFRRLTPTEVKDYLAAIA
Especificaciones
AAH47697 | |
proteasome (prosome, macropain) subunit, alpha type, 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMA2 | |
GST |
5683 | |
Wheat germ expression system | |
25 μg | |
HC3, MU, PMSA2, PSC2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |