missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23929-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.Especificaciones
PRPF6 | |
Polyclonal | |
Unconjugated | |
PRPF6 | |
ANT 1, C20orf14, hPrp6, Pre mRNA processing factor 6, PRP6 homolog, PRPF6, TOM, U5 102 kDa protein, U5 102K | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24148, 68879 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O94906, Q91YR7 | |
Prpf6 | |
PRPF6 Fusion Protein Ag13817 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |