Learn More
Invitrogen™ Human PRKACA (PKA), His Tag Recombinant Protein
Human PRKACA (PKA), His Tag Recombinant Protein
Marca: Invitrogen™ PR3791B
Detalles adicionales : Peso : 0.00900kg
Descripción
PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.
Especificaciones
NP_002721.1 | |
43.5 kDa | |
PRKACA (PKA) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: AGC Group | |
E. coli | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
PKA Kinase Family | |
Recombinant | |
Storage Buffer: 30mM KPO4, pH 7.5, 50% glycerol, 150mM KCl, 1mM EDTA, 1mM DTT Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.