missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRDX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10703-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to PRDX4.
Probably involved in redox regulation of the cell. Regulates the activation of NF-kappa-B in the cytosol by a modulation of I-kappa-B-alpha phosphorylation.Especificaciones
PRDX4 | |
Polyclonal | |
Unconjugated | |
PRDX4 | |
Antioxidant enzyme AOE372, AOE37 2, peroxiredoxin 4, Peroxiredoxin IV, PRDX4, PRX 4, Prx IV, Thioredoxin peroxidase AO372 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10549, 53381, 85274 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08807, Q13162, Q9Z0V5 | |
Prdx4 | |
PRDX4 Fusion Protein Ag1177 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |