missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPFIA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11492-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The PPFIA3 protein is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family, a family of transmembrane protein tyrosine phosphatases known to be important for axon guidance and mammary gland development. Similar to the other liprin family members, PPFIA3 can interact with ERC2, a member of the ELKS-Rab6-interacting protein-CAST family of proteins at the presynaptic termini, suggesting that these proteins may play a role in the molecular organization of presynaptic zones.Especificaciones
PPFIA3 | |
Polyclonal | |
Unconjugated | |
PPFIA3 | |
KIAA0654, Liprin alpha 3, LPNA3, PPFIA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140591, 76787, 8541 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75145, P60469, Q91Z79 | |
PPFIA3 | |
PPFIA3 Fusion Protein Ag1971 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |