missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PPARG (Human) Recombinant Protein
Human PPARG full-length ORF ( AAH06811, 1 a.a. - 477 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005468-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTMVDTEMPFWPTNFGISSVDLSVMEDHSHSFDIKPFTTVDFSSISTPHYEDIPFTRTDPVVADYKYDLKLQEYQSAIKVEPASPPYYSEKTQLYNKPHEEPSNSLMAIECRVCGDKASGFHYGVHACEGCKGFFRRTIRLKLIYDRCDLNCRIHKKSRNKCQYCRFQKCLAVGMSHNAIRFGRMPQAEKEKLLAEISSDIDQLNPESADLRALAKHLYDSYIKSFPLTKAKARAILTGKTTDKSPFVIYDMNSLMMGEDKIKFKHITPLQEQSKEVAIRIFQGCQFRSVEAVQEITGYAKSTPGFVNLDLNDQVTLLKYGVHEIIYTMLASLMNKDGVLISEGQGFMTREFLKSLRKPFGDFMEPKFEFAVKFNALELDDSDLAIFIAVIILSGDRPGLLNVKPIEDIQDNLLQALELQLKLNHPESSQLFAKLLQKMTDLRQIVTEHVQLLQVIKKTETDMSLHPLLQEIYKDLY
Especificaciones
AAH06811 | |
peroxisome proliferator-activated receptor gamma | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PPARG | |
GST |
5468 | |
Wheat germ expression system | |
25 μg | |
CIMT1, NR1C3, PPARG1, PPARG2, PPARgamma | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |