missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Cintas de etiquetado de un solo color de calidad industrial PowerMark™
Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.
Marca: Brady™ 013595
Detalles adicionales : Peso : 0.57830kg
Descripción
- Brady's BBP85 and PowerMark™ industrial grade print ribbon is super-tough and smear-proof to ensure your printed text last for years outdoors and in industrial areas.
Especificaciones
Cinta de etiquetado de un solo color | |
BBP85 Industrial Sign and Label Printer, PowerMark Sign and Label Printer, PowerMark Industrial Sign and Label Printer | |
6.22 in. | |
158 mm | |
Industrial | |
El texto impreso dura años a la intemperie y en zonas industriales |
Rojo | |
65.6 yd. | |
60 m | |
60 m x 158 mm | |
Lado de tinta dentro | |
1 rollo |