Learn More
Camlab™ Polypropylene Storage Box
Marca: Camlab™ RTP72051N
Detalles adicionales : Peso : 0.09000kg
Especificaciones
Natural | |
Cryo Storage Box | |
0.2 to 1 mL Tubes Up to 30 mm Diameter | |
5 x 10 |
Polypropylene | |
0.2 to 1mL Tubes Up to 30mm In Diameter | |
5 Pack |