Learn More
Camlab™ Gradilla de polipropileno
Marca: Camlab™ RTP711096N
Detalles adicionales : Peso : 0.26000kg
Especificaciones
Rack | |
Natural | |
Polypropylene | |
Laboratory Sample Storage | |
1 Each | |
198 mm | |
21 mm |
Lid | |
Tubes (96) | |
Yes | |
Freezer storage system | |
-40°C to +70°C | |
129 mm |