missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR1E Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16145-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae. Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. This gene encodes a member of the golgin family of proteins which are localized to the Golgi. Its encoded protein has been postulated to play a role in nuclear transport and Golgi apparatus localization. Several alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.Especificaciones
POLR1E | |
Polyclonal | |
Unconjugated | |
POLR1E | |
PAF53, POLR1E, PRAF1, RNA polymerase I subunit A49, RP11 405L18.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313245, 64424, 64425 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K202, Q9GZS1 | |
Polr1e | |
POLR1E Fusion Protein Ag9269 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |