missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PMS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18164-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to PMS2. The calculated molecular weight of PMS2 is 96 kDa, but the modified PMS2 is about 117 kDa.
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.Especificaciones
PMS2 | |
Polyclonal | |
Unconjugated | |
PMS2 | |
HNPCC4, PMS1 protein homolog 2, PMS2, PMS2CL, PMSL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18861, 288479, 5395 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54278, P54279 | |
Pms2 | |
PMS2 Fusion Protein Ag12651 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |