missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLXNB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 23795-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.Especificaciones
PLXNB1 | |
Polyclonal | |
Unconjugated | |
PLXNB1 | |
KIAA0407, plexin B1, PLXN5, PLXNB1, Semaphorin receptor SEP, SEP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235611, 5364 | |
-20°C | |
Liquid |
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43157, Q8CJH3 | |
PLXNB1 | |
PLXNB1 Fusion Protein Ag20719 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |