Learn More
Abnova™ PLEKHC1 Recombinant Protein
Human PLEKHC1 full-length ORF recombinant protein with GST-tag at N-terminal
Marca: Abnova™ H00010979-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Theoretical MW (kDa):100.54
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MALDGIRMPDGCYADGTWELSVHVTDLNRDVTLRVTGEVHIGGVMLKLVEKLDVKKDWSDHALWWEKKRTWLLKTHWTLDKYGIQADAKLQFTPQHKLLRLQLPNMKYVKVKVNFSDRVFKAVSDICKTFNIRHPEELSLLKKPRDPTKKKKKKLDDQSEDEALELEGPLITPGSGSIYSSPGLYSKTMTPTYDAHDGSPLSPTSAWFGDSALSEGNPGILAVSQPITSPEILAKMFKPQALLDKAKINQGWLDSSRSLMEQDVKENEALLLRFKYYSFFDLNPKYDAIRINQLYEQAKWAILLEEIECTEEEMMMFAALQYHINKLSIMTSENHLNNSDKEVDEVDAALSDLEITLEGGKTSTILGDITSIPELADYIKVFKPKKLTLKGYKQYWCTFKDTSISCYKSKEESSGTPAHQMNLRGCEVTPDVNISGQKFNIKLLIPVAEGMNEIWLRCDNEKQYAHWMAACRLASKGKTMADSSYNLEVQNILSSLKMQHLNPDPQLIPEQITTDITPECLVSPRYLKKYKNKQITARILEAHQNVAQMSLIEAKMRFIQAWQSLPEFGITHFIARFQGGKKEELIGIAYNRLIRMDASTGDAIKTWRFSNMKQWNVNWEIKMVTVEFADEVRLSFICTEVDCKVVHEFIGGYIFLSTRAKDQNESLDEEMFYKLTSGWV
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH17327 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
100.5 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MALDGIRMPDGCYADGTWELSVHVTDLNRDVTLRVTGEVHIGGVMLKLVEKLDVKKDWSDHALWWEKKRTWLLKTHWTLDKYGIQADAKLQFTPQHKLLRLQLPNMKYVKVKVNFSDRVFKAVSDICKTFNIRHPEELSLLKKPRDPTKKKKKKLDDQSEDEALELEGPLITPGSGSIYSSPGLYSKTMTPTYDAHDGSPLSPTSAWFGDSALSEGNPGILAVSQPITSPEILAKMFKPQALLDKAKINQGWLDSSRSLMEQDVKENEALLLRFKYYSFFDLNPKYDAIRINQLYEQAKWAILLEEIECTEEEMMMFAALQYHINKLSIMTSENHLNNSDKEVDEVDAALSDLEITLEGGKTSTILGDITSIPELADYIKVFKPKKLTLKGYKQYWCTFKDTSISCYKSKEESSGTPAHQMNLRGCEVTPDVNISGQKFNIKLLIPVAEGMNEIWLRCDNEKQYAHWMAACRLASKGKTMADSSYNLEVQNILSSLKMQHLNPDPQLIPEQITTDITPECLVSPRYLKKYKNKQITARILEAHQNVAQMSLIEAKMRFIQAWQSLPEFGITHFIARFQGGKKEELIGIAYNRLIRMDASTGDAIKTWRFSNMKQWNVNWEIKMVTVEFADEVRLSFICTEVDCKVVHEFIGGYIFLSTRAKDQNESLDEEMFYKLTSGWV | |
DKFZp686G11125/FLJ34213/FLJ44462/KIND2/MIG2/PLEKHC1/UNC112/UNC112B/mig-2 | |
FERMT2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10979 | |
PLEKHC1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FERMT2 | |
Human | |
Recombinant | |
Solution |