missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLCB3 Mouse anti-Human, Mouse, Rat, Clone: 1B8B3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66668-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.Especificaciones
PLCB3 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51432, Q01970, Q99JE6 | |
Plcb3 | |
PLCB3 Fusion Protein Ag15845 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1B8B3 | |
Unconjugated | |
PLCB3 | |
FLJ37084, Phospholipase C beta 3, PLC beta 3, PLCB3 | |
Mouse | |
Protein G | |
RUO | |
18797, 29322, 5331 | |
-20°C | |
Liquid |