missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAA Mouse anti-Human, Mouse, Rat, Clone: 5C10G9, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60244-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Phospholipase A-2-activating protein (PLAA), also named PLAP, can interact with ubiquitin and is involved in the maintece of ubiquitin levels. PLAA contains multiple consensus domains including WD repeats, PFU domain and PUL domain. Although the function of PLAA remains unclear, ubiquitin binding of PLAA might be the central role that connects ubiquitination and degradation in ERAD. Recent finding revealed that PLAA served as a novel nociceptive mediator after incision, and its expression level is regulated by miR-203.Especificaciones
PLAA | |
Monoclonal | |
1.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27612, P54319, Q9Y263 | |
Plaa | |
PLAA Fusion Protein Ag3224 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
5C10G9 | |
Unconjugated | |
PLAA | |
DOA1, PLA2P, PLAA, PLAP | |
Mouse | |
Protein A | |
RUO | |
116645, 18786, 9373 | |
-20°C | |
Liquid |