missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC zeta Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10860-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a constitutive kinase activity which is independent of calcium, and PKC activators, phosphatidylserine and diacylglycerol. Furthermore, it is insensitive to PKC inhibitors and cannot be activated by phorbol ester. The structural and biochemical properties indicate that the zeta subspecies is related to, but distinct from other isoenzymes of PKC.Especificaciones
PKC zeta | |
Polyclonal | |
Unconjugated | |
PRKCZ | |
PKC ZETA, PKC2, PRKCZ, Protein kinase C zeta type, protein kinase C, zeta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18762, 5590 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q02956, Q05513 | |
Prkcz | |
PKC zeta Fusion Protein Ag1310 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |