missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITPNA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16613-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of a family of lipid-binding proteins that transfer molecules of phosphatidylinositol or phosphatidylcholine between membrane surfaces. The protein is implicated in phospholipase C signaling and in the production of phosphatidylinositol 3,4,5-trisphosphate (PIP3) by phosphoinositide-3-kinase.Especificaciones
PITPNA | |
Polyclonal | |
Unconjugated | |
PITPNA | |
PI TP alpha, PITPN, PITPNA, PtdIns transfer protein alpha, PtdInsTP alpha, VIB1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18738, 29525, 5306 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P16446, P53810, Q00169 | |
Pitpna | |
PITPNA Fusion Protein Ag9849 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |