Learn More
Thermo Scientific™ Fosfato, solución de tampón de 0,2 M, pH 8,5
Used in molecular biology, biochemistry, and chromatography. It is used in the purification of antibodies.
Marca: Thermo Scientific™ J61925.AP
Detalles adicionales : CAS : 7558-80-7 Peso : 0.00050kg
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Especificaciones
Solución de tampón de 0,2 m | |
500mL | |
7558-80-7 | |
Líquido |
119,976 | |
Frasco de plástico | |
Temperaturas ambientes |