Learn More
PGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15161-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Phosphoglucomutases.Especificaciones
PGM1 | |
Polyclonal | |
Unconjugated | |
PGM1 | |
Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24645, 5236, 66681 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P36871, P38652, Q9D0F9 | |
PGM1 | |
PGM1 Fusion Protein Ag7269 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |