missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PGK2 (Human) Recombinant Protein
Human PGK2 full-length ORF ( AAH38843, 1 a.a. - 417 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005232-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSLSKKLTLDKLDVRGKRVIMRVDFNVPMKKNQITNNQRIKASIPSIKYCLDNGAKAVVLMSHLGRPDGVPMPDKYSLAPVAVELKSLLGKDVLFLKDCVGAEVEKACANPAPGSVILLENLRFHVEEEGKGQDPSGKKIKAEPDKIEAFRASLSKLGDVYVNDAFGTAHRAHSSMVGVNLPHKASGFLMKKELDYFAKALENPVRPFLAILGGAKVADKIQLIKNMLDKVNEMIIGGGMAYTFLKVLNNMEIGASLFDEEGAKIVKDIMAKAQKNGVRITFPVDFVTGDKFDENAQVGKATVASGISPGWMGLDCGPESNKNHAQVVAQARLIVWNGPLGVFEWDAFAKGTKALMDEIVKATSKGCITVIGGGDTATCCAKWNTEDKVSHVSTGGGASLELLEGKILPGVEALSNM
Especificaciones
AAH38843 | |
phosphoglycerate kinase 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PGK2 | |
GST |
5232 | |
Wheat germ expression system | |
10 μg | |
PGK-2, PGKB, PGKPS, dJ417L20.2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |