missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PERLD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17112-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Involved in the lipid remodeling steps of GPI-anchor maturation. Lipid remodeling steps consist in the generation of 2 saturated fatty chains at the sn-2 position of GPI-anchors proteins. Required for phospholipase A2 activity that removes an acyl-chain at the sn-2 position of GPI-anchors during the remodeling of GPI (Probable).Especificaciones
PERLD1 | |
Polyclonal | |
Unconjugated | |
PGAP3 | |
AGLA546, CAB2, COS16 homolog, hCOS16, per1 like domain containing 1, PERLD1, PGAP3, PP1498 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
93210 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96FM1 | |
PGAP3 | |
PERLD1 Fusion Protein Ag10796 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |