missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEDF Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26045-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The PEDF gene is a member of the serpin gene family. Serpins are a group of serine protease inhibitors, some of which have also been reported to exhibit neurotrophic activity.Especificaciones
PEDF | |
Polyclonal | |
Unconjugated | |
SERPINF1 | |
EPC 1, PEDF, PIG35, Serpin F1, SERPINF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5176 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36955 | |
SERPINF1 | |
PEDF Fusion Protein Ag23296 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |