missing translation for 'onlineSavingsMsg'
Learn More

PDGFRβ Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-1692-P

Detalles adicionales : Peso : 0.00100kg

 Ver más versiones de este producto

Código de producto. 12653137

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and β coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, α while PDGF BB can bind and activate to both PDGFR, α and β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Near C-terminal

Immunogen: A synthetic peptide derived from near the C-terminus of human PDGFRβ

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

PDGFR beta Ab-1
Polyclonal
1mg/mL
Rabbit
1 mL
Growth Factor
Human
IgG
Immunohistochemistry (Paraffin)
Polyclonal
Unconjugated
A synthetic peptide derived from near the C-terminus of human PDGFR, beta
RUO
Primary
Concentrated
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales