missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCSK9 Monoclonal Antibody (15), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543397
Detalles adicionales : Peso : 0.01000kg
Descripción
Specificity: Human PCSK9
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.Especificaciones
PCSK9 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8NBP7 | |
PCSK9 | |
Recombinant Human PCSK9 Protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
15 | |
Unconjugated | |
PCSK9 | |
AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
Mouse | |
Protein A | |
RUO | |
255738 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |