missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PCMT1 (Human) Recombinant Protein
Human PCMT1 full-length ORF ( AAH07501.1, 1 a.a. - 228 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005110-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAWKSGGASHSELIHNLRKNGIIKTDKVFEVMLATDRSHYAKCNPYMDSPQSIGFQATISAPHMHAYALELLFDQLHEGAKALDVGSGSGILTACFARMVGCTGKVIGIDHIKELVDDSVNNVRKDDPTLLSSGRVQLVVGDGRMGYAEEAPYDAIHVGAAAPVVPQALIDQLKPGGRLILPVGPAGGNQMLEQYDKLQDGSIKMKPLMGVIYVPLTDKEKQWSRDEL
Especificaciones
AAH07501.1 | |
protein-L-isoaspartate (D-aspartate) O-methyltransferase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5110 | |
Wheat germ expression system | |
25 μg | |
PCMT1 | |
GST |