missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCGF6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24103-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene contains a RING finger motif, which is most closely related to those of polycomb group (PcG) proteins RNF110/MEL-18 and BMI1. PcG proteins are known to form protein complexes and function as transcription repressors. This protein has been shown to interact with some PcG proteins and act as a transcription repressor. The activity of this protein is found to be regulated by cell cycle dependent phosphorylation. Alternatively spliced transcript variants encoding different isoforms have been identified.Especificaciones
PCGF6 | |
Polyclonal | |
Unconjugated | |
PCGF6 | |
MBLR, PCGF6, polycomb group ring finger 6, RING finger protein 134, RNF134 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84108 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BYE7 | |
PCGF6 | |
PCGF6 Fusion Protein Ag21124 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |