missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PANK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11768-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
PANK1 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA.Especificaciones
PANK1 | |
Polyclonal | |
Unconjugated | |
PANK1 | |
hPanK, hPanK1, PANK, PANK1, PANK1a, PANK1b, pantothenate kinase 1, Pantothenic acid kinase 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294088, 53354, 75735 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K4K6, Q8TE04 | |
PANK1 | |
PANK1 Fusion Protein Ag2324 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |