missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan Ras Mouse anti-Human, Mouse, Pig, Rat, CoraLite 555, Clone: 4H4G7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL555-60309-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
pan Ras Monoclonal antibody specifically detects pan Ras in Human, Mouse, Pig, Rat samples. It is validated for FC (Intra)Especificaciones
pan Ras | |
Monoclonal | |
1000 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
Mouse | |
20 μL | |
3845 | |
Store at -20°C | |
Liquid |
Flow Cytometry | |
4H4G7 | |
CoraLite 555 | |
BC011731 | |
Protein G purification | |
Primary | |
Human, Mouse, Pig, Rat | |
Antibody | |
IgG1 |