missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK6 Recombinant Human Protein
PAK6 Recombinant Human Protein
Marca: Invitrogen PV3502
Detalles adicionales : Peso : 0.00900kg
Descripción
PAK6 is a serine⁄threonine kinase expressed in the testis and prostate. It associates with both the estrogen and androgen receptors.Especificaciones
NP_064553 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
79.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PAK6, PAK5 | |
PAK6 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μM PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PAK6 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.25mg/mL | |
56924 | |
PAK6 Recombinant Human Protein | |
–80°C | |
1 Each | |
PAK6 Recombinant Human Protein | |
STE20 | |
221nmoles of phosphate transferred to PAKtide peptide substrate (CRRKSLVGpTPYWMAPE) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1.67μg/mL | |
His Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.