missing translation for 'onlineSavingsMsg'
Learn More

PAK4/6/7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 12460-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16820184

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 20-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The antibody can recognize PAK4/6/7. PAK4/6/7 belongs to the protein kinase superfamily, STE Ser/Thr protein kinase family and STE20 subfamily.

PAK4 is highly expressed in prostate, testis and colon. PAK4 interacts tightly with GTP-bound but not GDP-bound CDC42 and weakly with RAC. PAK4 phosphorylates and autophosphorylates and also activates the JNK pathway. PAK5, a member of the PAK family of protein kinases contains a CDC42/Rac1 interactive binding (CRIB) motif at the N-terminus and a Ste20-like kinase domain at the C-terminus. PAK5 preferentially binds to CDC42 in the presence of GTP and the CRIB motif is essential for this interaction. PAK6 protein cotranslocates into the nucleus with androgen receptor, which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development, in response to androgen. PAK6 was initially identified as an androgen receptor in a yeast two hybrid screen and was found to be highly expressed in testis and prostate tissues. Later experiments have shown it to be activated by MAP kinase 6 and p38 MAP kinase, suggesting that PAK6 may play a role in the cellular response to stress-related signals.
Especificaciones

Especificaciones

PAK4/6/7
Polyclonal
Unconjugated
PAK4
KIAA1264, p21 activated kinase 5, p21 activated kinase 7, PAK 5, PAK 7, PAK5, PAK7
Rabbit
Antigen Affinity Chromatography
RUO
10298, 214230, 241656, 292756, 296078, 311450, 56924, 57144, 70584
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O96013, Q3ULB5, Q8BTW9, Q8C015, Q9NQU5, Q9P286
Pak4, PAK5, PAK6, Pak7
PAK4/6/7 Fusion Protein Ag3127
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales