Learn More
p53 Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Achieve accurate, reproducible results with the Epredia™ p53 Ab-2, Mouse Monoclonal Antibody.
Marca: Epredia MS-105-P1ABX
Detalles adicionales : Peso : 1.00000kg
Descripción
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to mdm2, SV40 T antigen and human papilloma virus E6 protein p53 senses DNA damage and possibly facilitating repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.Host Species: Mouse
Clone: PAb 1801
Isotype: IgG1
Species Reactivity: Human specific.
Epitope: aa 32-79
Immunogen: Recombinant human p53 protein
Molecular Weight: 53kDa
Positive Control: SKBR-3 cells, MDA-231 cells
Cellular Localization: Nuclear
Recommended for:
- Western Blotting
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Especificaciones
p53 Ab-2 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cell Cycle | |
Human | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Western Blot | |
PAb 1801 | |
Unconjugated | |
Recombinant Human p53 protein | |
RUO | |
Primary | |
Concentrated |