missing translation for 'onlineSavingsMsg'
Learn More

P3H2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15723-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16815034

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 16-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the prolyl 3-hydroxylase subfamily of 2-oxo-glutarate-dependent dioxygenases. These enzymes play a critical role in collagen chain assembly, stability and cross-linking by catalyzing post-translational 3-hydroxylation of proline residues. Mutations in this gene are associated with nonsyndromic severe myopia with cataract and vitreoretinal degeneration, and downregulation of this gene may play a role in breast cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Especificaciones

Especificaciones

P3H2
Polyclonal
Unconjugated
P3H2
leprecan like 1, Leprecan like protein 1, LEPREL1, MLAT4, P3H2, Prolyl 3 hydroxylase 2
Rabbit
Antigen Affinity Chromatography
RUO
210530, 55214
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CG71, Q8IVL5
P3H2
P3H2 Fusion Protein Ag8444
20 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales